missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Calbiochem™ MACERASE™ Pectinase, Rhizopus sp.

Catalog No. 44120110KU
Change view
Click to view available options
Quantity:
10 kU
50 kU
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
44120110KU 10 kU
44120150KU 50 kU
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 44120110KU Supplier MilliporeSigma™ Supplier No. 44120110KU

Useful in isolating intact cells and protoplasts from higher plants

Note: 1KU = 1000 units.

Specifications

Color Pale Amber
Content And Storage 2°C to 8°C
Source Rhizopus sp.
Quantity 10 kU
Product Type Pectinase
Form Solid

For Research Use Only