missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Calbiochem™ Lysozyme, Human Neutrophil

Catalog No. 4403451MG
Change view
Click to view available options
Quantity:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
4403451MG 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 4403451MG Supplier MilliporeSigma™ Supplier No. 4403451MG

Takes part in antibacterial activity of leukocytes

  • Native lysozyme from human neutrophils
  • Occurs in specific granules of human polymorphonuclear leukocytes
  • Hydrolyzes β-1,4 linkages between N-acetylmuramic acid (2-acetamido-2-deoxy-3-O-[1′-carboxymethyl]-glucopyranose) residues in chitin

Specifications

Content And Storage -20°C
Source Human Neutrophil
Quantity 1 mg
Product Type Lysozyme, Human Neutrophil
Form Lyophilized

For Research use Only