missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Calbiochem™ Lectin, PHA-L, Phaseolus vulgaris

Catalog No. 4317845MG
Change view
Click to view available options
Quantity:
5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
4317845MG 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 4317845MG Supplier MilliporeSigma™ Supplier No. 4317845MG

Useful as specific marker in anterograde neuronal tracing

  • PHA-L is potent lymphocyte mitogen with narrow range
  • Requires calcium and manganese ions for activity

Specifications

Content And Storage -20°C
Quantity 5 mg
Product Type Lectin, PHA-L, Phaseolus vulgaris
Form Lyophilized

For Research use Only