missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Calbiochem™ GMBS

Catalog No. 442630100MG
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
442630100MG 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 442630100MG Supplier MilliporeSigma™ Supplier No. 442630100MG

Used for immobilizing enzymes on solid supports.

EMD Millipore Calbiochem™ GMBS is heterobifunctional reagent useful for preparation of conjugates such as secondary antibodies β-galactosidase for EIA procedures.

  • Heterobifunctional reagent
  • Preparation of conjugates
  • Immobilizing enzymes
  • Assay: ≥90% by HPLC
  • CAS: 80307-12-6
  • Molecular Formula: C12H12N2O6
  • Packaging: Glass Bottle

Recommended Storage

−20°C

Specifications

Description GMBS
Quantity 100 mg