missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MilliporeSigma™ Calbiochem™ Glutaraldehyde, 25% Aqueous Solution
Used for cross-linking proteins polyhydroxy materials.
Supplier: MilliporeSigma™ 354400500ML
Description
EMD Millipore Calbiochem™ Glutaraldehyde, 25% Aqueous Solution used as fixative for tissues.
- Used as cross-linking agent fixative
- Assay: ≥98% by TLC
- CAS: 111-30-8
- Color: Clear light yellow
Recommended Storage
15° to 30°C
Specifications
| Glutaraldehyde, 25% Aqueous Solution | |
| 25% aqueous solution | |
| 500 mL |