Learn More
MilliporeSigma™ Calbiochem™ FragEL™ TdT Enzyme DNA Fragmentation Detection Kit, Colorimetric
Description
- Non-isotopic kit
- Allows recognition of apoptotic nuclei in paraffin-embedded tissue sections, tissue cryosections, or in cell preparations fixed on slide by Fragment End Labeling (FragEL) of DNA
- Counterstaining with methyl green aids in the morphological evaluation and characterization of normal and apoptotic cells
Specifications
Specifications
| Content And Storage | -20°C |
| Quantity | 1 Each |
| Product Type | DNA Fragmentation Detection Kit |
For Research use Only