Learn More
MilliporeSigma™ Calbiochem™ FITC Labeling Kit
Catalog No. 3432101KIT
Change view
Click to view available options
Quantity:
1 Kit
Catalog No. Quantity
3432101KIT 1 Kit
1 options
Catalog No. 3432101KIT Supplier MilliporeSigma™ Supplier No. 3432101KIT
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Includes
Fluorescein (five vials), Carbonate buffer Concentrate, DMF Solvent, PBS buffer Concentrate and user protocol
Labels antibodies or proteins with fluorescein.
EMD Millipore Calbiochem™ FITC Labeling Kit is sufficient to perform at least five conjugations on proteins.
  • Direct labelling of primary antibody
  • Lower background overall higher signal-to-noise ratio
Recommended Storage
2° to 8°C
Specifications
Description FITC Labeling Kit
Kit Contents Fluorescein (five vials), Carbonate Buffer Concentrate, DMF Solvent, PBS Buffer Concentrate, and user protocol
Quantity 1 Kit