missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Calbiochem™ FITC, Isomer I

Catalog No. 343211GM
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
343211GM 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 343211GM Supplier MilliporeSigma™ Supplier No. 343211GM

Used to label antibodies or proteins.

EMD Millipore Calbiochem™ FITC, Isomer I is popular derivative of fluorescein.

  • Hygroscopic in nature
  • Derivative of fluorescein
  • Assay: ≥95% by TLC
  • CAS: 3326-32-7
  • Molecular Formula: C21H11NO5S
  • Packaging: Glass Bottle
  • Color: Orange Red

Recommended Storage

2° to 8°C

Specifications

Description FITC, Isomer I
Quantity 1 g