missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Calbiochem™ EMPIGEN™ BB Detergent, 30% Solution

Catalog No. 324690100ML
Change view
Click to view available options
Quantity:
100mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
324690100ML 100mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 324690100ML Supplier MilliporeSigma™ Supplier No. 324690100ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Can be used as detergent for solubilization of membrane components

  • Zwitterionic detergent
  • Useful in preparation of viral antigens, solubilization of keratins

Specifications

Content And Storage 15° to 30°C
Product Type BB Detergent, 30% Solution
Quantity 100mL

For Research Use Only