missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Calbiochem EGTA/AM

Catalog No. 32462810MG
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
32462810MG 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 32462810MG Supplier MilliporeSigma™ Supplier No. 32462810MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Application in Life Science research

Membrane-permeable form of the Ca2+-chelating agent EGTA

Specifications

Membrane Permeable Y
Purity ≥95% by HPLC
CAS 99590-86-0
Content And Storage -20°C
Form Oil
Quantity 10 mg
Synonym Ethyleneglycol-bis(β-aminoethyl)-N,N,Nʹ,Nʹ-tetraacetoxymethyl Ester
Solubility DMF, DMSO
Product Type Calbiochem EGTA/AM