missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Calbiochem™ EDTA, Disodium Salt, Dihydrate, Molecular Biology Grade

Catalog No. 3245031KG
Change view
Click to view available options
Quantity:
1 kg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
3245031KG 1 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 3245031KG Supplier MilliporeSigma™ Supplier No. 3245031KG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Application in Molecular Biology

Contaminants: DNases, proteases, RNases: none detected

Specifications

CAS 6381-92-6
Content And Storage 15°C to 30°C
Form Crystalline Solid
Quantity 1 kg
Synonym Ethylenediaminetetraacetic Acid, 2Na
Solubility Water
Product Type Calbiochem EDTA