missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Calbiochem™ EDTA, 0.5 M, pH 8.0, Molecular Biology Grade, DEPC-Treated

Catalog No. 324506100ML
Change view
Click to view available options
Quantity:
100 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
324506100ML 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 324506100ML Supplier MilliporeSigma™ Supplier No. 324506100ML

Suitable for use in molecular biology applications

Sterile-filtered solution of 0.5M EDTA in H2O treated with diethyl pyrocarbonate (DEPC)

Specifications

CAS 150-38-9
Content And Storage 15°C to 30°C
Form Liquid
Quantity 100 mL
Synonym Ethylenediaminetetraacetic Acid Trisodium Salt
Product Type Calbiochem EDTA