missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Calbiochem™ DSP, Water-Soluble

Catalog No. 322133100MG
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
322133100MG 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 322133100MG Supplier MilliporeSigma™ Supplier No. 322133100MG

Used to immobilize proteins on supports containing amino groups.

EMD Millipore Calbiochem™ DSP, Water-Soluble is bifunctional cross-linker.

  • Disulfide bond cleaved with 1% NaBH4, 50mM DTT, or 100mM β-mercaptoethanol
  • Packaging: Glass Bottle
  • Color: Off-White to Light Tan

Recommended Storage

2° to 8°C

Specifications

Description DSP, Water-Soluble
Quantity 100 mg