missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Calbiochem™ Deoxyribonucleic Acid, Sodium Salt, Salmon Testes

Catalog No. p-7060202
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
2620125GM 5 g
2620121GM 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 2620125GM Supplier MilliporeSigma™ Supplier No. 2620125GM
Only null left

Purified DNA from salmon testes

  • Purified DNA from salmon testes
  • N/P ratio: 1.5-1.8
  • Absorption (0.001% solution in phosphate buffer, pH 7.0, 260 nm): ≥0.155

Specifications

Buffer Phosphate Buffer
CAS 9007-49-2
Color Off-white
Content And Storage Store at +2°C to +8°C
pH 7.0
Quantity 5 g
Form Solid