missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Calbiochem™ D(+)Melezitose, Monohydrate

Catalog No. 444201100GM
Change view
Click to view available options
Quantity:
100 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
444201100GM 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 444201100GM Supplier MilliporeSigma™ Supplier No. 444201100GM

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Substrate for α-glucosidase

A substrate for α-glucosidase

Specifications

CAS 10030-67-8
Color White
Content And Storage Store at +15°C to +30°C
Formula Weight 522.5g/mol
Molecular Formula C18H32O16 · H2O
Purity ≥98% by HPLC
Quantity 100 g
Synonym GlcαFruβ2,1Glcα, H2O
Form Solid