missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Calbiochem™ D-Mannose

Catalog No. 4440100GM
Change view
Click to view available options
Quantity:
100 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
4440100GM 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 4440100GM Supplier MilliporeSigma™ Supplier No. 4440100GM

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

For Life Science reasearch application

D-Mannose - CAS 3458-28-4 - Calbiochem

Specifications

CAS 3458-28-4
Color White
Content And Storage Store at +15°C to +30°C
Formula Weight 180.2g/mol
Molecular Formula C6H12O6
Purity ≥98% by GC and HPLC
Quantity 100 g
Form Solid