missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Calbiochem™ α-D-Glucose-6-phosphate, Monosodium Salt

Catalog No. 3467645GM
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
3467645GM 5 g
3467641GM 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 3467645GM Supplier MilliporeSigma™ Supplier No. 3467645GM

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Aqueous stock solutions are stable for up to 6 weeks at -20°C

α-D-Glucose-6-phosphate, Monosodium Salt - CAS 54010-71-8 - Calbiochem

Specifications

CAS 54010-71-8
Content And Storage Store at +2°C to +8°C
Formula Weight 282.1g/mol
Molecular Formula C6H12NaO9P
Purity >99% by enzymatic assay
Quantity 5 g
Form Crystalline Solid