missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Calbiochem™ D(+)Glucosamine, Hydrochloride

Catalog No. 346299100GM
Change view
Click to view available options
Quantity:
100 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
346299100GM 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 346299100GM Supplier MilliporeSigma™ Supplier No. 346299100GM

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Aminosugar

D(+)Glucosamine, Hydrochloride - CAS 66-84-2 - Calbiochem

Specifications

CAS 66-84-2
Color White
Content And Storage Store at +15°C to +30°C
Formula Weight 215.6g/mol
Molecular Formula C6H13NO5 ·HCl
Purity ≥99% by Assay
Quantity 100 g
Synonym 2-Amino-2-deoxy-D-glucose, Chitosamine
Form Solid