missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Calbiochem™ D(+)Galactosamine, Hydrochloride

Catalog No. 345391GM
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
345391GM 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 345391GM Supplier MilliporeSigma™ Supplier No. 345391GM

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Powerful inhibitor of hepatic RNA synthesis

  • A powerful inhibitor of hepatic RNA synthesis
  • Increases the adherence of polymorphonuclear leukocytes to hepatic endothelial cells and induces superoxide production
  • Induces hepatic damage, causing macrophage infiltration

Specifications

CAS 1772-03-8
Color White
Content And Storage Store at +15°C to +30°C
Formula Weight 215.6g/mol
For Use With (Application) Inhibitor of hepatic RNA synthesis
Purity ≥98% by titration
Quantity 1 g
Form Solid