missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Calbiochem™ D-Fructose-6-phosphate, Disodium Salt

Catalog No. 3443421GM
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
3443421GM 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 3443421GM Supplier MilliporeSigma™ Supplier No. 3443421GM

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Glycolytic intermediate

A glycolytic intermediate formed by the isomerization of glucose-6-phosphate

Specifications

CAS 26177-86-6
Color White
Content And Storage Store at +2°C to +8°C
Formula Weight 304.1g/mol
Purity ≥95% by chromatography
Quantity 1 g
Form Solid