missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Calbiochem™ Coelenterazine

Catalog No. 233900250UG
Change view
Click to view available options
Quantity:
250 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
233900250UG 250 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 233900250UG Supplier MilliporeSigma™ Supplier No. 233900250UG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

A cell-permeable aequorin luminophore that acts as a very sensitive and specific chemiluminescence probe of the superoxide anion.

Can also be used for detecting changes in intracellular Ca2+ in cells that have been transfected with apoaequorin cDNA. Also acts as a powerful antioxidant.

Specifications

Content And Storage Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C
Quantity 250 μg
Form Solid