missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Calbiochem™ Cleland’s Reagent, ULTROL™ Grade

Catalog No. 2331531GM
Change view
Click to view available options
Quantity:
1 g
5 g
10 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
2331531GM 1 g
2331535GM 5 g
23315310GM 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 2331531GM Supplier MilliporeSigma™ Supplier No. 2331531GM

Protective agent for SH groups

  • Absorbance: (100mM, 280nm): ≤0.05
  • Oxidized dithiothreitol: ≤0.5%

Specifications

Molecular Weight (g/mol) 154.24
Purity ≥99% by titration
CAS 12-3-3483
Molecular Formula C4H10O2S2
Content And Storage 2°C to 8°C
Form Solid
Quantity 1 g
Synonym Dithiothreitol, DTT
Solubility EtOH, Water
Product Type Calbiochem Cleland’s Reagent