missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MilliporeSigma™ Calbiochem™ Cleland’s Reagent, Molecular Biology Grade
Description
- Contaminants: DNases, proteases, RNases: none detected
- Absorbance (100mM, 1cm, 260nm): ≤0.500; (100mM, 1cm, 280nm): ≤0.100
- Heavy metals: ≤0.001%
Specifications
Specifications
| Molecular Weight (g/mol) | 154.24 |
| Purity | ≥99% (Titration) |
| Molecular Formula | C4H10O2S2 |
| Content And Storage | 2°C to 8°C |
| Form | Crystalline Solid |
| Quantity | 10 g |
| Synonym | 1,4-Dithiothreitol, DTT |
| Solubility | EtOH, Water |
| Product Type | Calbiochem Cleland’s Reagent |