missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Calbiochem™ Cleland’s Reagent, Molecular Biology Grade

Catalog No. 23315610GM
Change view
Click to view available options
Quantity:
1 g
10 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
23315610GM 10 g
2331561GM 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 23315610GM Supplier MilliporeSigma™ Supplier No. 23315610GM

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Protective agent for SH groups

  • Contaminants: DNases, proteases, RNases: none detected
  • Absorbance (100mM, 1cm, 260nm): ≤0.500; (100mM, 1cm, 280nm): ≤0.100
  • Heavy metals: ≤0.001%

Specifications

Molecular Weight (g/mol) 154.24
Purity ≥99% (Titration)
Molecular Formula C4H10O2S2
Content And Storage 2°C to 8°C
Form Crystalline Solid
Quantity 10 g
Synonym 1,4-Dithiothreitol, DTT
Solubility EtOH, Water
Product Type Calbiochem Cleland’s Reagent