missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Calbiochem™ Cerulenin, Cephalosporium caerulens

Catalog No. 2195575MG
Change view
Click to view available options
Quantity:
5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
2195575MG 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 2195575MG Supplier MilliporeSigma™ Supplier No. 2195575MG

Inhibits sterol fatty acid biosynthesis.

EMD Millipore Calbiochem™ Cerulenin, Cephalosporium caerulens inhibits HMG-CoA synthase activity.

  • Antifungal antibiotic
  • Binds in equimolar ratio to β-keto-acyl-ACP synthase
  • Inhibit feeding induce dramatic weight loss in mice
  • Assay: ≥98% by TLC
  • CAS: 17397-89-6
  • Molecular Formula: C12H17NO3
  • Color: White to Off-White

Specifications

Quantity 5 mg
Content And Storage 2°C to 8°C