Learn More
MilliporeSigma™ Calbiochem™ Caspase-8 Detection Kit (FITC-IETD-FMK)
Convenient, and sensitive method to measure caspase-8 in living cells
Supplier:  MilliporeSigma™ QIA1131KIT
Catalog No. QIA1131KIT
Only null left
Description
Description
  • Fluorescent marker, FITC-IETD-FMK, is labeled, cell-permeable, non-toxic inhibitor
  • Binds irreversibly to activated caspase-8 in living cells
  • Detected by measurement of fluorescence in fluorescent microplate reader, flow cytometer, or by fluorescence microscopy
Specifications
Specifications
1 Kit
Caspase-8 Detection Kit
Product Suggestions
Product Suggestions