Learn More
MilliporeSigma™ Calbiochem™ Caspase-3 Detection Kit (FITC-DEVD-FMK)
Catalog No. QIA911KIT
Change view
Click to view available options
Quantity:
1 Kit (100 tests)
Catalog No. Quantity
QIA911KIT 1 Kit (100 tests)
1 options
Catalog No. QIA911KIT Supplier MilliporeSigma™ Supplier No. QIA911KIT
For detection of activated caspase-3 in living cells
  • Cell-permeable, non-toxic inhibitor that binds irreversibly to activated caspase-3 in apoptotic cells
  • Can be measured by fluorescence microscopy, fluorescence plate reader, or flow cytometry
Specifications
Content And Storage -20°C
Quantity 1 Kit (100 tests)
Product Type Caspase-3 Detection Kit (FITC-DEVD-FMK)
For Research use Only