missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Calbiochem™ Caffeic Acid

Catalog No. 205546500MG
Change view
Click to view available options
Quantity:
500 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
205546500MG 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 205546500MG Supplier MilliporeSigma™ Supplier No. 205546500MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Reported to have anti-carcinogenic anti-inflammatory properties.

EMD Millipore Caffeic Acid is dietary compound which is cell-permeable, selective, non-competitive inhibitor of 5-lipoxygenase.

  • Inhibits Cu2+-induced LDL oxidation in initiation phase
  • Acts as prooxidant in propagation phase
  • An irreversible inhibitor of glutathione S-transferases
  • Non-competitive inhibitor of xanthine oxidase
  • Assay: ≥95% by HPLC
  • CAS: 331-39-5
  • Molecular Formula: C9H8O4
  • Color: Tan

Specifications

Quantity 500 mg
Content And Storage 2°C to 8°C