missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MilliporeSigma™ Calbiochem™ BQ-123, Sodium Salt
Description
Primary target Ki: 40nM for ETA versus 2.5μM for ETB
Specifications
Specifications
| Color | White |
| Content And Storage | Store at −20°C |
| Formula Weight | 632.7g/mol |
| Formulation | Supplied as a sodium salt. Sold on the basis of peptide content |
| Molecular Formula | C31H41N6O7 · Na |
| Purity | ≥95% by HPLC |
| Quantity | 5 mg |
| Target | ETA (endothelin) receptor antagonist |
| Form | Solid |