missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Calbiochem™ BQ-123, Sodium Salt

Catalog No. 052338311MG
Change view
Click to view available options
Quantity:
5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
052338311MG 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 052338311MG Supplier MilliporeSigma™ Supplier No. 052338311MG

Highly potent, selective, and competitive ETA receptor antagonist

Primary target Ki: 40nM for ETA versus 2.5μM for ETB

Specifications

Color White
Content And Storage Store at −20°C
Formula Weight 632.7g/mol
Formulation Supplied as a sodium salt. Sold on the basis of peptide content
Molecular Formula C31H41N6O7 · Na
Purity ≥95% by HPLC
Quantity 5 mg
Target ETA (endothelin) receptor antagonist
Form Solid