missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Calbiochem™ Cleland’s Reagent

Catalog No. 2331551GM
Change view
Click to view available options
Quantity:
1 g
10 g
25 g
100 g
500 g
1 kg
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
2331551GM 1 g
23315510GM 10 g
23315525GM 25 g
233155100GM 100 g
233155500GM 500 g
2331551KG 1 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 2331551GM Supplier MilliporeSigma™ Supplier No. 2331551GM

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Protective agent for SH groups

  • Cyclizes as it reduces disulfides to thiols, so reaction is "driven" to completion
  • Does not contain waxy surfactant found in many other preparations
  • Oxidized dithiothreitol: ≤0.5%

Specifications

Molecular Weight (g/mol) 154.24
Purity ≥97% (Titration)
Molecular Formula C4H10O2S2
Content And Storage 2°C to 8°C
Form Powder
Quantity 1 g
Synonym DTT, Dithiothreitol
Solubility EtOH, Water
Product Type Calbiochem Cleland’s Reagent