missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Calbiochem™ Y-27632

Catalog No. 6880001MG
Change view
Click to view available options
Quantity:
1 mg
5 mg
10 mg
100 mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
6880001MG 1 mg
6880005MG 5 mg
68800010MG 10 mg
688000100MG 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 6880001MG Supplier MilliporeSigma™ Supplier No. 6880001MG

Y-27632A, CAS 146986-50-7, is a cell-permeable, reversible, inhibitor of Rho kinases (Ki = 140 nM for p160ROCK). Enhances survival & cloning efficiency of ESC without affecting their pluripotency.

  • Packaged Under Inert Gas
  • Cell Permeable

Specifications

Color White
Purity 95% by HPLC
Solubility H2O
Molecular Formula C14H21N3O·2HCl·H2O
CAS 146986-50-7
Content And Storage Hygroscopic
Protect from Light
Protein p160 Rho-associated protein kinases (ROCK)
Quantity 1 mg
Molecular Weight (g/mol) 338.3g/mol
Form Solid