missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MilliporeSigma™ Calbiochem™ Y-27632
Y-27632A, CAS 146986-50-7, is a cell-permeable, reversible, inhibitor of Rho kinases (Ki = 140 nM for p160ROCK). Enhances survival & cloning efficiency of ESC without affecting their pluripotency.
Supplier: MilliporeSigma™ 68800010MG
Description
- Packaged Under Inert Gas
- Cell Permeable
Chemical Identifiers
| White | |
| Hygroscopic Protect from Light |
|
| Solid |
| 95% by HPLC | |
| 338.3g/mol |
Specifications
| White | |
| Hygroscopic Protect from Light |
|
| Y-27632 | |
| Solid |
| 95% by HPLC | |
| 10 mg | |
| 338.3g/mol |