missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Calbiochem™ Adenosine 3'-Phosphate 5'-Phosphosulfate, Tetralithium Salt

Catalog No. 11841010MG
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11841010MG 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11841010MG Supplier MilliporeSigma™ Supplier No. 11841010MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Sulfate donor substrate for sulfotransferases, including Sulfotransferase

  • Prepared enzymatically
  • A sulfate donor substrate for sulfotransferases, including Sulfotransferase, Nod Factor
  • Unstable at room temperature

Specifications

CAS 102029-54-9
Color White
Content And Storage Store at ≤ -70°C
Formula Weight 31g/mol
Molecular Formula C10H11N5O13P2S · 4Li
Purity ≥80% by HPLC
Quantity 10 mg
Synonym PAPS, 4Li
Form Solid