missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Calbiochem™ 5'-Deoxy-5'-methylthioadenosine

Catalog No. 26058550MG
Change view
Click to view available options
Quantity:
50 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
26058550MG 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 26058550MG Supplier MilliporeSigma™ Supplier No. 26058550MG

Inhibits protein carboxymethyltransferase, induces apoptosis in leukemia U937 cells.

EMD Millipore Calbiochem™ 5'-Deoxy-5'-methylthioadenosine also inhibits FGF-2 receptor tyrosine kinase activity.

  • Cell-permeable, reversible, ATP-competitive naturally-occurring co-product of polyamine biosynthesis that acts as an endogenous substrate of methylthioadenosine phosphorylase (MTAP)
  • Inhibits proliferation of human astrocytes, glioma cells
  • Assay: ≥99% by HPLC
  • CAS: 2457-80-9
  • Molecular Formula: C11H15N5O3S
  • Color: White

Specifications

Quantity 50 mg
Content And Storage -20°C