missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Calbiochem™ 2-Deoxy-D-glucose

Catalog No. 259721GM
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
259721GM 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 259721GM Supplier MilliporeSigma™ Supplier No. 259721GM

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Reported to reduce the incidence of doxorubicin-induced apoptosis

  • Non-metabolizable derivative of glucose that can be carried by the glucose carrier in human erythrocytes
  • An indirect inhibitor of hexokinase
  • Reported to reduce the incidence of doxorubicin-induced apoptosis

Specifications

CAS 154-17-6
Color Off-white to white
Content And Storage Store at +2°C to +8°C
Formula Weight 164.2g/mol
Molecular Formula C6H12O5
Purity ≥98% by HPLC
Quantity 1 g
Form Powder