Learn More
Specifications
| Chemical Name or Material | Cal-Ex™ Decalcifier |
|---|---|
| Physical Form | Liquid |
| Boiling Point | 100°C |
| CAS | 6381-92-6 , 7647-01-0 , 7732-18-5 |
| pH | 2.0 |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity & Availability | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity & Availability | ||||
|
CS5101D
|
Fisher Chemical
CS510 1 GH |
1 L | Poly Bottle |
Each for $125.52
Case of 6 Each for $753.12
|
|
||||
|
CS51010D
|
Fisher Chemical
CS510 10 GH |
10 L | PolyPac™ |
Each for $292.46
|
|
||||
Safety and Handling
DANGER!
Emergency Overview
Causes burns by all exposure routes. Use only under a chemical fume hood. Wash off immediately with plenty of water for at least 15 minutes. Immediate medical attention is required. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Immediate medical attention is required. Move to fresh air. If breathing is difficult, give oxygen. Do not use mouth-to-mouth resuscitation if victim ingested or inhaled the substance; induce artifici. Immediate medical attention is required. Do not induce vomiting. Call a physician or Poison Control Center immediately. Call a physician or Poison Control Center immediately. .
NFPA
Health:3
Flammability:0
Instability:0
DOTInformation : DOT Class 8, : Corrosive