missing translation for 'onlineSavingsMsg'
Learn More

STERIS™ Cage-Klenz 250 Acid-Based Detergent

Cage-Klenz 250 Detergent: Safe, effective removal of hard water scale, urine scale and organic residues in research animal caging. Phosphate-free, citric acid-based formula. Efficacy, materials compatibility, cost savings, and environmentally friendly. pH (1%): 2.6.

Specifications

Product Type Detergent
Color-coded No
Form Clear Liquid
pH 2.6 (Diluted)
Scent Unscented
View More Specs

Products 3
Catalog Number Mfr. No. Size Container Type Price Quantity & Availability  
Catalog Number Mfr. No. Size Container Type Price Quantity & Availability  
19999642
View Documents Promotion Details
STERIS™
1K2505
5 gal. Pail N/A
Only null left
 
19999641
View Documents Promotion Details
STERIS™
1K2502
30 gal. Drum N/A
Only null left
 
19999643
View Documents Promotion Details
STERIS™
1K25D9
200 gal. Tote N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

Cage-Klenz 250 Detergent is specially formulated for the safe and effective removal of hard water scale, urine scale and organic residues found in research animal caging.

HOW CAGE-KLENZ 250 DETERGENT WORKS

Cage-Klenz 250 Detergent is a liquid phosphate-free, citric acid-based detergent.

WHY CAGE-KLENZ 250 DETERGENT?

  • Efficacy - Effectively cleans hard water and urine scale in both soft and hard water
  • Flexibility - Excellent materials compatibility with a wide variety of caging materials including polycarbonate
  • Cost Savings - Reduces utility consumption and cleaning time due with low foaming, free-rinsing formulation
  • Environmentally Friendly - phosphate-free formulation with biodegradable surfactants
  • Reliability - Manufactured in a cGMP compliant, FDA registered and ISO 9001 certified facility

Physical Properties: pH (1%): 2.6, typical

Specifications

Specifications

Detergent
Clear Liquid
Unscented
No
2.6 (Diluted)
Non-sterile
Videos
SDS
Documents

Documents