missing translation for 'onlineSavingsMsg'
Learn More

STERIS™ Cage-Klenz 250 Acid-Based Detergent

Catalog No. 19999643
Change view
Click to view available options
Size:
5 gal.
30 gal.
200 gal.
Container Type:
Drum
Pail
Tote
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size Container Type
19999643 200 gal. Tote
19999642 5 gal. Pail
19999641 30 gal. Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 19999643 Supplier STERIS™ Supplier No. 1K25D9

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Cage-Klenz 250 Detergent: Safe, effective removal of hard water scale, urine scale and organic residues in research animal caging. Phosphate-free, citric acid-based formula. Efficacy, materials compatibility, cost savings, and environmentally friendly. pH (1%): 2.6.

Cage-Klenz 250 Detergent is specially formulated for the safe and effective removal of hard water scale, urine scale and organic residues found in research animal caging.

HOW CAGE-KLENZ 250 DETERGENT WORKS

Cage-Klenz 250 Detergent is a liquid phosphate-free, citric acid-based detergent.

WHY CAGE-KLENZ 250 DETERGENT?

  • Efficacy - Effectively cleans hard water and urine scale in both soft and hard water
  • Flexibility - Excellent materials compatibility with a wide variety of caging materials including polycarbonate
  • Cost Savings - Reduces utility consumption and cleaning time due with low foaming, free-rinsing formulation
  • Environmentally Friendly - phosphate-free formulation with biodegradable surfactants
  • Reliability - Manufactured in a cGMP compliant, FDA registered and ISO 9001 certified facility

Physical Properties: pH (1%): 2.6, typical

Specifications

Product Type Detergent
Color-coded No
Form Clear Liquid
pH 2.6 (Diluted)
Scent Unscented
Sterility Non-sterile
Container Type Tote
Size 200 gal.