missing translation for 'onlineSavingsMsg'
Learn More
Learn More
STERIS™ Cage-Klenz 250 Acid-Based Detergent
Description
Cage-Klenz 250 Detergent is specially formulated for the safe and effective removal of hard water scale, urine scale and organic residues found in research animal caging.
HOW CAGE-KLENZ 250 DETERGENT WORKS
Cage-Klenz 250 Detergent is a liquid phosphate-free, citric acid-based detergent.
WHY CAGE-KLENZ 250 DETERGENT?
- Efficacy - Effectively cleans hard water and urine scale in both soft and hard water
- Flexibility - Excellent materials compatibility with a wide variety of caging materials including polycarbonate
- Cost Savings - Reduces utility consumption and cleaning time due with low foaming, free-rinsing formulation
- Environmentally Friendly - phosphate-free formulation with biodegradable surfactants
- Reliability - Manufactured in a cGMP compliant, FDA registered and ISO 9001 certified facility
Physical Properties: pH (1%): 2.6, typical
Specifications
Specifications
| Product Type | Detergent |
| Color-coded | No |
| Form | Clear Liquid |
| pH | 2.6 (Diluted) |
| Scent | Unscented |
| Sterility | Non-sterile |
| Container Type | Drum |
| Size | 30 gal. |