missing translation for 'onlineSavingsMsg'
Learn More

Caffeic Acid 98.0+%, TCI America™

Supplier:  TCI America C00025G

 View more versions of this product

Catalog No. C00025G


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

Caffeic Acid
Yellow
MFCD00004392
caffeic acid, 3,4-dihydroxycinnamic acid, 3-3,4-dihydroxyphenyl acrylic acid, trans-caffeate, 3,4-dihydroxy-trans-cinnamate, 2e-3-3,4-dihydroxyphenyl prop-2-enoic acid, 3,4-dihydroxybenzeneacrylic acid, e-3-3,4-dihydroxyphenyl acrylic acid, trans-caffeic acid, 3-3,4-dihydroxyphenyl propenoic acid
C1=CC(=C(C=C1C=CC(=O)O)O)O
180.159
CHEBI:16433
≥98.0% (HPLC,T)
331-39-5
C9H8O4
5 g
QAIPRVGONGVQAS-DUXPYHPUSA-N
(E)-3-(3,4-dihydroxyphenyl)prop-2-enoic acid
689043
180.16
Crystalline Powder

Chemical Identifiers

331-39-5
180.159
QAIPRVGONGVQAS-DUXPYHPUSA-N
689043
(E)-3-(3,4-dihydroxyphenyl)prop-2-enoic acid
C9H8O4
MFCD00004392
caffeic acid, 3,4-dihydroxycinnamic acid, 3-3,4-dihydroxyphenyl acrylic acid, trans-caffeate, 3,4-dihydroxy-trans-cinnamate, 2e-3-3,4-dihydroxyphenyl prop-2-enoic acid, 3,4-dihydroxybenzeneacrylic acid, e-3-3,4-dihydroxyphenyl acrylic acid, trans-caffeic acid, 3-3,4-dihydroxyphenyl propenoic acid
CHEBI:16433
C1=CC(=C(C=C1C=CC(=O)O)O)O
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

RTECSNumber : GD8950000

TSCA : No

SDS
Documents

Documents

Promotions

Promotions