missing translation for 'onlineSavingsMsg'
Learn More

Caffeic Acid 98.0+%, TCI America™

Catalog No. p-7124167
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C00025G 5 g
C000225G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C00025G Supplier TCI America Supplier No. C00025G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 331-39-5
Molecular Formula C9H8O4
Molecular Weight (g/mol) 180.159
MDL Number MFCD00004392
InChI Key QAIPRVGONGVQAS-DUXPYHPUSA-N
Synonym caffeic acid, 3,4-dihydroxycinnamic acid, 3-3,4-dihydroxyphenyl acrylic acid, trans-caffeate, 3,4-dihydroxy-trans-cinnamate, 2e-3-3,4-dihydroxyphenyl prop-2-enoic acid, 3,4-dihydroxybenzeneacrylic acid, e-3-3,4-dihydroxyphenyl acrylic acid, trans-caffeic acid, 3-3,4-dihydroxyphenyl propenoic acid
PubChem CID 689043
ChEBI CHEBI:16433
IUPAC Name (E)-3-(3,4-dihydroxyphenyl)prop-2-enoic acid
SMILES C1=CC(=C(C=C1C=CC(=O)O)O)O

Specifications

Color Yellow
Quantity 5 g
Formula Weight 180.16
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Caffeic Acid
RTECSNumber GD8950000
TSCA No