missing translation for 'onlineSavingsMsg'
Learn More

Cadmium Sulfate, 0.05% (w/v), Ricca Chemical

Catalog No. R1707500500
Change view
Click to view available options
Quantity:
500 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
R1707500500 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. R1707500500 Supplier Ricca Chemical Company Supplier No. R1707500500A

Chemical Identifiers

CAS 7790-84-3 , 7732-18-5

Specifications

Color Colorless
CAS Min % 98.0, 0.05
CAS Max % 100.0, 0.05
Percent Purity 0.05%
Grade Laboratory
Packaging Natural Poly Bottle
Quantity 500 mL
Physical Form Liquid
Concentration or Composition Notes 0.05% (w/v)
Chemical Name or Material Cadmium Sulfate