Learn More
Cadmium Standard For ICP, 1000μg/mL in 2% HNO3, Assurance Grade, SPEX CertiPrep™
ISO/IEC Guide 34 Certified, NIST-Traceable Certified Reference Material
Supplier: SPEX CertiPrep Group LLC PLCD22T
Includes: NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet
Description
Inorganic Standard manufactured for AA, ICP, ICP/MS, IC, XRF, and other analytical instrumentation
| Analyte | Concentration |
|---|---|
| Cd | 1000μg/mL |
Certifications
Accredited by A2LA for ISO/IEC 17025 and ISO/IEC Guide 34 and certified by UL-DQS to ISO 9001
Compliance
GHS/OSHA Complaint
Specifications
| Cadmium Standard | |
| 7440-43-9,7697-37-2,7732-18-5 | |
| Up to 68 elements are scanned with found values for ICP and ICP-MS standards. The found value of the trace metallic impurities is reported on the Certificate of Analysis. | |
| 0.1,2.0,97.9 | |
| MFCD00010914 | |
| AA, ICP, ICP/MS, IC, XRF, and Other Analytical Instrumentation | |
| BDOSMKKIYDKNTQ-UHFFFAOYSA-N | |
| 2% HNO3 | |
| 112.41 | |
| 23973 | |
| Assurance |
| Triple-leached LDPE bottle | |
| Assurance Certificate of Analysis reports up to 30 elements at ppb levels. | |
| 0.1,2.0,97.9 | |
| Cd | |
| AA, ICP | |
| Certified Reference Material | |
| [Cd] | |
| cadmium | |
| 250 mL | |
| CHEBI:22977 | |
| Matrix Concentration Cat. No. 2-5% HNO3 1000μg/mLPLCD2-2Y, -2T, -2X 10,000 PLCD2-3Y, -3X 2-5% HCl 1000μg/mL PLCD1-2X |
Chemical Identifiers
| 7440-43-9 | |
| 112.41 | |
| BDOSMKKIYDKNTQ-UHFFFAOYSA-N | |
| CHEBI:22977 | |
| [Cd] |
| Cd | |
| MFCD00010914 | |
| 23973 | |
| cadmium |
Safety and Handling
Causes skin irritation, serious eye irritation. May cause cancer. Suspected of damaging fertility or the unborn child.
DOTInformation : Hazardous
Recommended Storage : Ambient
Product intended for laboratory use only