Learn More
Cadmium, plasma standard solution, Specpure™ Cd 10,000μg/mL
CAS: 7697-37-2 | HNO3 | 63.01 g/mol
$225.96 - $526.05
Chemical Identifiers
| CAS | 7697-37-2 |
|---|---|
| Molecular Formula | HNO3 |
| Molecular Weight (g/mol) | 63.01 |
| MDL Number | MFCD00011349 |
| InChI Key | GRYLNZFGIOXLOG-UHFFFAOYSA-N |
| PubChem CID | 944 |
| ChEBI | CHEBI:48107 |
| IUPAC Name | nitric acid |
| SMILES | O[N+]([O-])=O |
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 7697-37-2 | |
| 63.01 | |
| GRYLNZFGIOXLOG-UHFFFAOYSA-N | |
| CHEBI:48107 | |
| O[N+]([O-])=O |
| HNO3 | |
| MFCD00011349 | |
| 944 | |
| nitric acid |
Specifications
| 7697-37-2,7732-18-5 | |
| Liquid | |
| MFCD00011349 | |
| GRYLNZFGIOXLOG-UHFFFAOYSA-N | |
| nitric acid | |
| 100 mL | |
| CHEBI:48107 | |
| Cadmium |
| 23 hPa (17mm Hg) at 20°C | |
| HNO3 | |
| Colorless | |
| O[N+]([O-])=O | |
| 63.01 | |
| 944 | |
| Nitric acid: 5.0%; Cadmium: 1.0%; Water: 94.0% |
Safety and Handling
P201-P202-P234-P260-P264b-P271-P281-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P390-P501c
H290-H314-H335-H350
DOTInformation : Hazard Class: 8, Packing Group: III
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only