missing translation for 'onlineSavingsMsg'
Learn More

Cadmium Carbonate, Technical, Spectrum™ Chemical

Catalog No. 18609399
Change view
Click to view available options
Quantity:
125 g
500 g
2.5 kg
45 kg
Packaging:
Poly Bottle
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18609399 125 g Poly Bottle
18609400 500 g Poly Bottle
18609397 2.5 kg Poly Bottle
18609398 45 kg Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 18609399 Supplier Spectrum Chemical Mfg Cor Supplier No. C1023125GM
Get Up to $500 in Chemicals with Purchase
View all promotions

Chemical Identifiers

CAS 513-78-0
Molecular Weight (g/mol) 172.41 g/mol

Specifications

CAS Max % 1
Grade Technical
Packaging Poly Bottle
Quantity 125 g
Chemical Name or Material Cadmium Carbonate

CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.