missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cadmium Carbonate, Powder, Reagent, 97%, Spectrum™ Chemical
C1020, 513-78-0, CdCO3
$2566.47 - $2566.47
Chemical Identifiers
| CAS | 513-78-0 |
|---|---|
| Molecular Formula | CCdO3 |
| Molecular Weight (g/mol) | 172.42 |
| MDL Number | MFCD00010918 |
| InChI Key | GKDXQAKPHKQZSC-UHFFFAOYSA-L |
| IUPAC Name | cadmium(2+) carbonate |
| SMILES | [Cd++].[O-]C([O-])=O |
Description
Spectrum™ Chemical Cadmium Carbonate, Powder, Reagent , also known as Otavite, a naturally occurring mineral. The Reagent grade denotes that this chemical is the highest quality commercially available and that the American Chemical Society has not officially set any specifications for this material. Spectrum Chemical manufactured Reagent grade products meet the toughest regulatory standards for quality and purity.Chemical Identifiers
| 513-78-0 | |
| 172.42 | |
| GKDXQAKPHKQZSC-UHFFFAOYSA-L | |
| [Cd++].[O-]C([O-])=O |
| CCdO3 | |
| MFCD00010918 | |
| cadmium(2+) carbonate |
Specifications
| 513-78-0 | |
| 500 g | |
| MFCD00010918 | |
| GKDXQAKPHKQZSC-UHFFFAOYSA-L | |
| cadmium(2+) carbonate | |
| 97% | |
| 25.5-27.5% |
| 100% | |
| CCdO3 | |
| UN2570 | |
| [Cd++].[O-]C([O-])=O | |
| 172.42 | |
| Reagent | |
| Poly Bottle |