missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cadmium Acetate Dihydrate, purum p.a., ≥98.0% (KT), Honeywell™
Purum p.a.,≥98.0%
$1070.79 - $1070.79
Chemical Identifiers
| CAS | 5743-04-4 |
|---|---|
| Molecular Formula | C4H10CdO6 |
| Molecular Weight (g/mol) | 266.53 |
| MDL Number | MFCD00150020 |
| InChI Key | AUIZLSZEDUYGDE-UHFFFAOYSA-L |
Description
- Products with a guarantee certificate and/or suitable for the stated analytical application.
Specifications
| 256°C | |
| Solid | |
| C4H10CdO6 | |
| MFCD00150020 | |
| 3730567 | |
| 266.53 | |
| ≥98.0% (KT) | |
| Poly Bottle | |
| Cadmium Acetate Dihydrate |
| 5743-04-4 | |
| 500 g | |
| Cd(CH3COO)2 · 2H2O | |
| UN3077 | |
| AUIZLSZEDUYGDE-UHFFFAOYSA-L | |
| 266.53g/mol | |
| purum p.a. | |
| 2.01g/cu.cm |
Safety and Handling
P201-P261-P273-P280-P308 + P313-P501
H302 + H312 + H332-H350-H410
DOTInformation : 9/PGIII
EINECSNumber : 208-853-2
RTECSNumber : AF7505000