missing translation for 'onlineSavingsMsg'
Learn More

Cadmium Acetate Dihydrate, purum p.a., ≥98.0% (KT), Solstice

Catalog No. 60045180
Change view
Click to view available options
Quantity:
500 g
Packaging:
Poly Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
60045180 500 g Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 60045180 Supplier Solstice Supplier No. 20901500G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Purum p.a.,≥98.0%

  • Products with a guarantee certificate and/or suitable for the stated analytical application.

Chemical Identifiers

CAS 5743-04-4
Molecular Formula C4H10CdO6
Molecular Weight (g/mol) 266.53
MDL Number MFCD00150020
InChI Key AUIZLSZEDUYGDE-UHFFFAOYSA-L

Specifications

Melting Point 256°C
Physical Form Solid
Quantity 500 g
Linear Formula Cd(CH3COO)2 · 2H2O
UN Number UN3077
Beilstein 3730567
Formula Weight 266.53g/mol
Percent Purity ≥98.0% (KT)
Grade purum p.a.
Packaging Poly Bottle
Density 2.01g/cu.cm
Chemical Name or Material Cadmium Acetate Dihydrate
Show More Show Less
Hazard Statement P201-P261-P273-P280-P308 + P313-P501
Precautionary Statement H302 + H312 + H332-H350-H410
DOTInformation 9/PGIII
EINECSNumber 208-853-2
RTECSNumber AF7505000