missing translation for 'onlineSavingsMsg'
Learn More

Cabozantinib (S)-malate, Thermo Scientific™

Catalog No. p-7238602
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC468902500 250 mg
AC468900010 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC468902500 Supplier Thermo Scientific Chemicals Supplier No. 468902500
Only null left

Chemical Identifiers

CAS 1140909-48-3
Molecular Formula C32H30FN3O10
Molecular Weight (g/mol) 635.60
MDL Number MFCD20923480
InChI Key HFCFMRYTXDINDK-UHFFFAOYNA-N
IUPAC Name 2-hydroxybutanedioic acid; N'1-{4-[(6,7-dimethoxyquinolin-4-yl)oxy]phenyl}-N1-(4-fluorophenyl)cyclopropane-1,1-dicarboxamide
SMILES OC(CC(O)=O)C(O)=O.COC1=C(OC)C=C2C(OC3=CC=C(NC(=O)C4(CC4)C(=O)NC4=CC=C(F)C=C4)C=C3)=CC=NC2=C1

Specifications

Packaging Glass Bottle
Certificate of Analysis Available
Physical Form Crystalline Powder
Color White to Light Beige
Melting Point 166°C to 172°C
Quantity 250 mg
Chemical Name or Material Cabozantinib (S)-malate
Hazard Category Exclamation mark
Hazard Statement Health hazard
Recommended Storage Normal conditions